accessibility menu, dialog, popup

UserName

CAS RN 53566-95-3

NOO

CAS RN 53566-95-3

IUPAC Name: 4-[(4-formylphenyl)(phenyl)amino]benzaldehyde
Synonyms: 4-[(4-formylphenyl)(phenyl)amino]benzaldehyde
Molecular Weight (g/mol): 301.35
Molecular Formula: C20H15NO2
InChi Key: DOUAFMIJGIUWJX-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results