missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-formylphenyl) Ether 98.0+%, TCI America™

Catalog No. B50991G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50991G 1 g
B50995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50991G Supplier TCI America Supplier No. B50991G

Chemical Identifiers

CAS 2215-76-1
Molecular Formula C14H10O3
Molecular Weight (g/mol) 226.23
MDL Number MFCD08276812
InChI Key GXZZHLULZRMUQC-UHFFFAOYSA-N
PubChem CID 11368119
IUPAC Name 4-(4-formylphenoxy)benzaldehyde
SMILES O=CC1=CC=C(OC2=CC=C(C=O)C=C2)C=C1

Specifications

Melting Point 66°C
Color White-Yellow
Boiling Point 160°C
Quantity 1 g
Synonym 4,4′C-Diformyldiphenyl Ether, 4,4′C-Oxydibenzaldehyde
Formula Weight 226.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-formylphenyl) Ether