missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl) disulfide, 98%

Catalog No. AAH3258903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3258903 1 g
AAH3258906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3258903 Supplier Thermo Scientific Chemicals Supplier No. H3258903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 405-31-2 | C12H8F2S2 | 254.313 g/mol

Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-fluorophenyl) disulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 405-31-2
Molecular Formula C12H8F2S2
Molecular Weight (g/mol) 254.313
MDL Number MFCD00816758
InChI Key SLXZJIKDNHDPKL-UHFFFAOYSA-N
Synonym di-4-fluorophenyl sulfide, bis 4-fluorophenyl disulfide, 1-fluoro-4-4-fluorophenyl disulfanyl benzene, 1,1'-disulfanediylbis 4-fluorobenzene, 4-fluorophenyldisulfide, 4-fluorophenyl disulfide, 4-fluorophenyl disulphide, di-p-fluorophenyl sulfide, bis 4-fluorophenyl persulfide, 1,2-bis 4-fluorophenyl disulfane
PubChem CID 136252
IUPAC Name 1-fluoro-4-[(4-fluorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1F)SSC2=CC=C(C=C2)F

Specifications

Boiling Point 141°C (1 mmHg)
Quantity 1 g
Solubility Information Soluble in methanol and acetone.
Formula Weight 254.31
Percent Purity 98%
Chemical Name or Material Bis(4-fluorophenyl) disulfide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only