missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-cyanophenyl) Ether 98.0+%, TCI America™

Catalog No. p-7119760
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27905G 5 g
B279025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27905G Supplier TCI America Supplier No. B27905G

Chemical Identifiers

CAS 6508-04-9
Molecular Formula C14H8N2O
Molecular Weight (g/mol) 220.23
InChI Key RSAUOQFEFINEDM-UHFFFAOYSA-N
PubChem CID 81004
IUPAC Name 4-(4-cyanophenoxy)benzonitrile
SMILES C1=CC(=CC=C1C#N)OC2=CC=C(C=C2)C#N

Specifications

Melting Point 184°C
Color White
Boiling Point 220°C
Quantity 5 g
UN Number 3439
Synonym 4,4′C-Dicyanodiphenyl Ether, 4,4′C-Oxydibenzonitrile
Formula Weight 220.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-cyanophenyl) Ether