Learn More
Bis(4-chlorophenyl) sulfone, 99%
CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol
Supplier: Thermo Scientific Chemicals A1893836
Description
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.
Solubility
Insoluble in water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents.
Specifications
| Bis(4-chlorophenyl) sulfone | |
| 147°C to 150°C | |
| 233°C (451°F) | |
| MFCD00000619 | |
| 2052955 | |
| Insoluble in water. | |
| ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1 | |
| 287.15 | |
| 287.17 |
| 80-07-9 | |
| 249°C to 251°C (10 mmHg) | |
| C12H8Cl2O2S | |
| 500 g | |
| 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone | |
| GPAPPPVRLPGFEQ-UHFFFAOYSA-N | |
| 1-chloro-4-(4-chlorobenzenesulfonyl)benzene | |
| 6625 | |
| 99% |
Chemical Identifiers
| 80-07-9 | |
| 287.15 | |
| GPAPPPVRLPGFEQ-UHFFFAOYSA-N | |
| 6625 |
| C12H8Cl2O2S | |
| MFCD00000619 | |
| 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone | |
| ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1 |
Safety and Handling
P264b-P280i-P305+P351+P338
H319-H500
EINECSNumber : 201-247-9
RTECSNumber : WR3450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only