missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorophenyl) sulfone, 99%

Catalog No. AAA1893822
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1893822 100 g
AAA1893836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1893822 Supplier Thermo Scientific Chemicals Supplier No. A1893822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 80-07-9 | C12H8Cl2O2S | 287.15 g/mol

Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4'-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(4-chlorophenyl) sulfone is used as a precursor to prepare 4,4′-diamino-diphenyl sulfone and polysulfone plastics. It is also used to prepare rigid and temperature resistant polymers such as PES or Udel through polymerization with bisphenol-A and bisphenol-S. Further, the udel polymer finds applications in plumbing pipes, printer cartridges and automobile fuses.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 80-07-9
Molecular Formula C12H8Cl2O2S
Molecular Weight (g/mol) 287.15
MDL Number MFCD00000619
InChI Key GPAPPPVRLPGFEQ-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl sulfone, bis 4-chlorophenyl sulfone, p,p'-dichlorodiphenyl sulfone, sulfone, bis p-chlorophenyl, 4-chlorophenyl sulfone, 1,1'-sulfonylbis 4-chlorobenzene, bis p-chlorophenyl sulfone, benzene, 1,1'-sulfonylbis 4-chloro, p-chlorophenyl sulfone, 4,4'-dichlorodiphenylsulfone
PubChem CID 6625
SMILES ClC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(Cl)C=C1

Specifications

Melting Point 147°C to 150°C
Boiling Point 249°C to 251°C (10 mmHg)
Flash Point 233°C (451°F)
Quantity 100 g
Beilstein 2052955
Solubility Information Insoluble in water.
IUPAC Name 1-chloro-4-(4-chlorophenyl)sulfonylbenzene
Formula Weight 287.17
Percent Purity 99%
Chemical Name or Material Bis(4-chlorophenyl) sulfone
Hazard Category H319-H500
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 201-247-9
RTECSNumber WR3450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only