missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-chlorophenyl) disulfide, 98+%

Catalog No. p-7028342
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2560406 5 g
AAB2560414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2560406 Supplier Thermo Scientific Chemicals Supplier No. B2560406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1142-19-4 | C12H8Cl2S2 | 287.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1142-19-4
Molecular Formula C12H8Cl2S2
Molecular Weight (g/mol) 287.216
MDL Number MFCD00013642
InChI Key ZIXXRXGPBFMPFD-UHFFFAOYSA-N
Synonym 4,4'-dichlorodiphenyl disulfide, bis 4-chlorophenyl disulfide, disulfide, bis 4-chlorophenyl, ddds, ddds pesticide, disulfide, bis p-chlorophenyl, p-chlorophenyl disulfide, 4-chlorophenyl disulfide, bis p-chlorophenyl disulfide, 4,4'-dichloro diphenyl disulfide
PubChem CID 14360
IUPAC Name 1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene
SMILES C1=CC(=CC=C1SSC2=CC=C(C=C2)Cl)Cl

Specifications

Melting Point 71°C to 74°C
Quantity 5 g
Beilstein 1644663
Formula Weight 287.23
Percent Purity ≥98%
Chemical Name or Material Bis(4-chlorophenyl) disulfide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-531-2
RTECSNumber JO0766000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only