accessibility menu, dialog, popup

UserName

CAS RN 1142-19-4

IUPAC Name: 1-chloro-4-[(4-chlorophenyl)disulfanyl]benzene
Synonyms: bis(4-chlorophenyl) disulfide
Molecular Weight (g/mol): 287.22
Molecular Formula: C12H8Cl2S2
InChi Key: ZIXXRXGPBFMPFD-UHFFFAOYSA-N
SMILES: ClC1=CC=C(SSC2=CC=C(Cl)C=C2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)