missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-bromophenyl) Sulfide 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 115°C |
| Color | White |
| Boiling Point | 210°C |
| Quantity | 5 g |
| Formula Weight | 344.06 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(4-bromophenyl) Sulfide |
Safety and Handling
| RTECSNumber | WP9900000 |
| TSCA | No |