missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl) Ether 97.0+%, TCI America™

Catalog No. B083425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B083425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B083425G Supplier TCI America Supplier No. B083425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2050-47-7
Molecular Formula C12H8Br2O
Molecular Weight (g/mol) 328.003
MDL Number MFCD00000095
InChI Key YAWIAFUBXXPJMQ-UHFFFAOYSA-N
Synonym bis 4-bromophenyl ether, 4,4'-dibromodiphenyl ether, 4,4'-oxybis bromobenzene, 4-bromophenyl ether, p,p'-dibromodiphenyl ether, usaf do-61, 1-bromo-4-4-bromophenoxy benzene, bis p-bromophenyl ether, ether, bis p-bromophenyl, benzene, 1,1'-oxybis 4-bromo
PubChem CID 16305
IUPAC Name 1-bromo-4-(4-bromophenoxy)benzene
SMILES C1=CC(=CC=C1OC2=CC=C(C=C2)Br)Br

Specifications

Melting Point 60°C
Boiling Point 205°C
Quantity 25 g
Formula Weight 328.00
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-bromophenyl) Ether
EINECSNumber (3)-0061
RTECSNumber KN0175000
TSCA Yes