missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-bromophenyl) ether, 99%

Catalog No. p-7049632
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1668406 5 g
AA1668414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1668406 Supplier Thermo Scientific Chemicals Supplier No. 01668406
Only null left

CAS: 2050-47-7 | C12H8Br2O | 328.003 g/mol

Bis-(4-bromophenyl)-ether has been used in the preparation of 4-[4-(diethoxyphosphoryl)phenoxy]phenyl-phosphonic acid diethyl ester. It is used for medicine and as organic intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2050-47-7
Molecular Formula C12H8Br2O
Molecular Weight (g/mol) 328.003
MDL Number MFCD00000095
InChI Key YAWIAFUBXXPJMQ-UHFFFAOYSA-N
Synonym bis 4-bromophenyl ether, 4,4'-dibromodiphenyl ether, 4,4'-oxybis bromobenzene, 4-bromophenyl ether, p,p'-dibromodiphenyl ether, usaf do-61, 1-bromo-4-4-bromophenoxy benzene, bis p-bromophenyl ether, ether, bis p-bromophenyl, benzene, 1,1'-oxybis 4-bromo
PubChem CID 16305
IUPAC Name 1-bromo-4-(4-bromophenoxy)benzene
SMILES C1=CC(=CC=C1OC2=CC=C(C=C2)Br)Br

Specifications

Melting Point 56°C to 60°C
Color White
Boiling Point 338°C to 340°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 328.01
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Bis(4-bromophenyl) ether
EINECSNumber 218-090-7
RTECSNumber KN0175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only