missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(4-bromophenyl)acetylene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 187°C |
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 336.03 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(4-bromophenyl)acetylene |
Safety and Handling
| TSCA | No |