missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-biphenylyl)amine 98.0+%, TCI America™

Catalog No. b24291g
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24291G 1 g
B24295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24291G Supplier TCI America Supplier No. B24291G
Only null left

Chemical Identifiers

CAS 102113-98-4
Molecular Formula C24H19N
Molecular Weight (g/mol) 321.42
MDL Number MFCD08276279
InChI Key JAUCIDPGGHZXRP-UHFFFAOYSA-N
PubChem CID 504244
IUPAC Name N-{[1,1'-biphenyl]-4-yl}-[1,1'-biphenyl]-4-amine
SMILES N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 1 g
Synonym 4,4′C-Iminobis(biphenyl), Di([1,1′C-biphenyl]-4-yl)amine
Formula Weight 321.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(4-biphenylyl)amine