accessibility menu, dialog, popup

UserName

HN

CAS RN 102113-98-4

IUPAC Name: N-{[1,1'-biphenyl]-4-yl}-[1,1'-biphenyl]-4-amine
Synonyms: N-{[1,1'-biphenyl]-4-yl}-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 321.42
Molecular Formula: C24H19N
InChi Key: JAUCIDPGGHZXRP-UHFFFAOYSA-N
SMILES: N(C1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results