missing translation for 'onlineSavingsMsg'
Learn More

Bis[4-(2-hydroxyethoxy)phenyl] Sulfone 97.0+%, TCI America™

Catalog No. b1590500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1590500G 500 g
B159025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1590500G Supplier TCI America Supplier No. B1590500G

Chemical Identifiers

CAS 27205-03-4
Molecular Formula C16H18O6S
Molecular Weight (g/mol) 338.374
MDL Number MFCD00130282
InChI Key UTNSTOOXQPHXJQ-UHFFFAOYSA-N
Synonym 4-(2-Hydroxyethoxy)phenyl Sulfone
PubChem CID 95071
IUPAC Name 2-[4-[4-(2-hydroxyethoxy)phenyl]sulfonylphenoxy]ethanol
SMILES C1=CC(=CC=C1OCCO)S(=O)(=O)C2=CC=C(C=C2)OCCO

Specifications

Melting Point 181°C
Color White
Quantity 500 g
Formula Weight 338.37
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[4-(2-hydroxyethoxy)phenyl] Sulfone
EINECSNumber (3)-3814
TSCA No