missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3-fluorophenyl) Disulfide 97.0+%, TCI America™
$46.60 - $136.08
Chemical Identifiers
| CAS | 63930-17-6 |
|---|---|
| Molecular Formula | C12H8F2S2 |
| Molecular Weight (g/mol) | 254.313 |
| MDL Number | MFCD01318104 |
| InChI Key | LFVFHTBHAIVNJQ-UHFFFAOYSA-N |
| Synonym | bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide |
| PubChem CID | 2737794 |
| IUPAC Name | 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene |
| SMILES | C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F |
Chemical Identifiers
| 63930-17-6 | |
| 254.313 | |
| LFVFHTBHAIVNJQ-UHFFFAOYSA-N | |
| 2737794 | |
| C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F |
| C12H8F2S2 | |
| MFCD01318104 | |
| bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide | |
| 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene |
Specifications
| 63930-17-6 | |
| C12H8F2S2 | |
| 1 g | |
| LFVFHTBHAIVNJQ-UHFFFAOYSA-N | |
| 1-fluoro-3-[(3-fluorophenyl)disulfanyl]benzene | |
| 2737794 | |
| ≥97.0% (GC) | |
| Bis(3-fluorophenyl) Disulfide |
| Yellow | |
| MFCD01318104 | |
| bis 3-fluorophenyl disulfide, bis 3-fluorophenyl disulphide, 1,2-bis 3-fluorophenyl disulfane, 3,3'-difluoro diphenyl disulfide, 1-fluoro-3-3-fluorophenyl disulfanyl benzene, 3,3'-difluorodiphenyl disulfide, bis-3-fluorophenyl disulfide, disulfide, bis 3-fluorophenyl, di3-fluorophenyl disulfide | |
| C1=CC(=CC(=C1)SSC2=CC=CC(=C2)F)F | |
| 254.313 | |
| 254.31 | |
| Liquid |
Safety and Handling
TSCA : No