missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(3-amino-4-hydroxyphenyl) Sulfone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 231°C |
| Quantity | 5 g |
| Synonym | 3,3′C-Diamino-4,4′C-dihydroxydiphenyl Sulfone |
| Formula Weight | 280.30 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis(3-amino-4-hydroxyphenyl) Sulfone |
Safety and Handling
| TSCA | No |