missing translation for 'onlineSavingsMsg'
Learn More

Bis(3-amino-4-hydroxyphenyl) Sulfone 98.0+%, TCI America™

Catalog No. B28595G
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B28595G 5 g
B285925G 25 g
2 options

Catalog No. B28595G

Supplier: TCI America B28595G

Only null left

Chemical Identifiers

CAS 7545-50-8
Molecular Formula C12H12N2O4S
Molecular Weight (g/mol) 280.298
InChI Key KECOIASOKMSRFT-UHFFFAOYSA-N
PubChem CID 82047
IUPAC Name 2-amino-4-(3-amino-4-hydroxyphenyl)sulfonylphenol
SMILES C1=CC(=C(C=C1S(=O)(=O)C2=CC(=C(C=C2)O)N)N)O

Specifications

Melting Point 231°C
Quantity 5 g
Synonym 3,3′C-Diamino-4,4′C-dihydroxydiphenyl Sulfone
Formula Weight 280.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(3-amino-4-hydroxyphenyl) Sulfone