missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl] Sulfone 96.0+%, TCI America™
$55.92 - $55.92
Chemical Identifiers
| CAS | 53714-39-9 |
|---|---|
| Molecular Formula | C16H14Br4O6S |
| Molecular Weight (g/mol) | 653.958 |
| MDL Number | MFCD00152428 |
| InChI Key | OFBQIWFJRDGFEK-UHFFFAOYSA-N |
| Synonym | Bis[4-(2-hydroxyethoxy)-3,5-dibromophenyl] Sulfone |
| PubChem CID | 104566 |
| IUPAC Name | 2-[2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]sulfonylphenoxy]ethanol |
| SMILES | C1=C(C=C(C(=C1Br)OCCO)Br)S(=O)(=O)C2=CC(=C(C(=C2)Br)OCCO)Br |
Chemical Identifiers
| 53714-39-9 | |
| 653.958 | |
| OFBQIWFJRDGFEK-UHFFFAOYSA-N | |
| 104566 | |
| C1=C(C=C(C(=C1Br)OCCO)Br)S(=O)(=O)C2=CC(=C(C(=C2)Br)OCCO)Br |
| C16H14Br4O6S | |
| MFCD00152428 | |
| Bis[4-(2-hydroxyethoxy)-3,5-dibromophenyl] Sulfone | |
| 2-[2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]sulfonylphenoxy]ethanol |
Specifications
| 53714-39-9 | |
| C16H14Br4O6S | |
| 25 g | |
| OFBQIWFJRDGFEK-UHFFFAOYSA-N | |
| 2-[2,6-dibromo-4-[3,5-dibromo-4-(2-hydroxyethoxy)phenyl]sulfonylphenoxy]ethanol | |
| 104566 | |
| ≥96.0% (GC) | |
| Bis[3,5-dibromo-4-(2-hydroxyethoxy)phenyl] Sulfone |
| White-Yellow | |
| MFCD00152428 | |
| Bis[4-(2-hydroxyethoxy)-3,5-dibromophenyl] Sulfone | |
| C1=C(C=C(C(=C1Br)OCCO)Br)S(=O)(=O)C2=CC(=C(C(=C2)Br)OCCO)Br | |
| 653.958 | |
| 653.96 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-3206
TSCA : No