missing translation for 'onlineSavingsMsg'
Learn More

Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate, 95%

Catalog No. p-7035659
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5337806 5 g
AAH5337814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5337806 Supplier Thermo Scientific Chemicals Supplier No. H5337806
Only null left

CAS: 75203-51-9 | C26H24Cl6O8 | 677.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 75203-51-9
Molecular Formula C26H24Cl6O8
Molecular Weight (g/mol) 677.17
MDL Number MFCD00012038
InChI Key PURKHUDOTFUVNG-UHFFFAOYSA-N
Synonym cppo, bis 2-carbopentyloxy-3,5,6-trichlorophenyl oxalate, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl oxalate, bis 2-carbopentyloxy-3,5,6-trichloro-phenyl oxal, ethanedioic acid, bis 2,3,5-trichloro-6-pentyloxy carbonyl phenyl ester, pentyl 3,4,6-trichloro-2-2-oxo-2-2,3,5-trichloro-6-pentyloxy carbonyl phenoxy acetyl oxy benzoate, bis 2,3,5-trichloro-6-pentoxycarbonylphenyl oxalate, bis 2,3,5-trichloro-6-pentoxycarbonyl-phenyl oxalate
PubChem CID 3018575
IUPAC Name bis(2,3,5-trichloro-6-pentoxycarbonylphenyl) oxalate
SMILES CCCCCOC(=O)C1=C(C(=C(C=C1Cl)Cl)Cl)OC(=O)C(=O)OC2=C(C(=CC(=C2Cl)Cl)Cl)C(=O)OCCCCC

Specifications

Melting Point 76°C to 80°C
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 677.19
Percent Purity 95%
Chemical Name or Material Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate
EINECSNumber 278-124-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only