missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 677.17 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Bis[3,4,6-trichloro-2-(pentyloxycarbonyl)phenyl] Oxalate |
Safety and Handling
| EINECSNumber | (3)-4136 |
| TSCA | Yes |