accessibility menu, dialog, popup

UserName

CH3OOClClClOOOOClClClOOCH3

CAS RN 30431-54-0

IUPAC Name: pentyl 2,3,5-trichloro-6-[(2-oxo-2-{3,4,6-trichloro-2-[(pentyloxy)carbonyl]phenoxy}acetyl)oxy]benzoate
Synonyms: pentyl 2,3,5-trichloro-6-[(2-oxo-2-{3,4,6-trichloro-2-[(pentyloxy)carbonyl]phenoxy}acetyl)oxy]benzoate
Molecular Weight (g/mol): 677.17
Molecular Formula: C26H24Cl6O8
InChi Key: TZZLVFUOAYMTHA-UHFFFAOYSA-N
SMILES: CCCCCOC(=O)C1=C(Cl)C(Cl)=CC(Cl)=C1OC(=O)C(=O)OC1=C(Cl)C=C(Cl)C(Cl)=C1C(=O)OCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results