missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-nitrophenyl) Disulfide 98.0+%, TCI America™

Catalog No. B050325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B050325G 25 g
B0503500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B050325G Supplier TCI America Supplier No. B050325G

Chemical Identifiers

CAS 1155-00-6
Molecular Formula C12H8N2O4S2
Molecular Weight (g/mol) 308.33
MDL Number MFCD00007130
InChI Key NXCKJENHTITELM-UHFFFAOYSA-N
Synonym bis 2-nitrophenyl disulfide, disulfide, bis 2-nitrophenyl, 2,2'-dinitrodiphenyl disulfide, o-nitrophenyl disulfide, 2-nitrophenyl disulfide, o,o'-dinitrodiphenyl disulfide, disulfide, bis o-nitrophenyl, bis o-nitrophenyl disulfide, 1,2-bis 2-nitrophenyl disulfane
PubChem CID 70861
IUPAC Name 1-nitro-2-[(2-nitrophenyl)disulfanyl]benzene
SMILES [O-][N+](=O)C1=CC=CC=C1SSC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 192°C
Quantity 25 g
Formula Weight 308.33
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-nitrophenyl) Disulfide
EINECSNumber (3)-1129
TSCA Yes