Learn More
Bis-(2-methylallyl)cycloocta-1,5-diene ruthenium(II) complex, 30-32% Ru, Thermo Scientific™
$117.59 - $396.66
Chemical Identifiers
| CAS | 12289-94-0 |
|---|---|
| Molecular Formula | C16H26Ru |
| Molecular Weight (g/mol) | 319.45 |
| MDL Number | MFCD00216965,MFCD00216965 |
| InChI Key | POYBJJLKGYXKJH-UHFFFAOYSA-N |
| PubChem CID | 91884701 |
| IUPAC Name | (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+) |
| SMILES | [Ru++].CC([CH2-])=C.CC([CH2-])=C.C1CC=CCCC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC295782500
|
Thermo Scientific Chemicals
295782500 |
250mg | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC295780010
|
Thermo Scientific Chemicals
295780010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 12289-94-0 | |
| 319.45 | |
| POYBJJLKGYXKJH-UHFFFAOYSA-N | |
| (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+) |
| C16H26Ru | |
| MFCD00216965,MFCD00216965 | |
| 91884701 | |
| [Ru++].CC([CH2-])=C.CC([CH2-])=C.C1CC=CCCC=C1 |
Specifications
| 12289-94-0 | |
| Glass bottle | |
| MFCD00216965,MFCD00216965 | |
| POYBJJLKGYXKJH-UHFFFAOYSA-N | |
| (5Z)-cycloocta-1,5-diene;2-methanidylprop-1-ene;ruthenium(2+) | |
| 91884701 | |
| 30-32% Ru |
| Authentic | |
| C16H26Ru | |
| 250mg | |
| [Ru++].CC([CH2-])=C.CC([CH2-])=C.C1CC=CCCC=C1 | |
| 319.45 | |
| 319.44 | |
| Bis-(2-methylallyl)cycloocta-1, 5-diene ruthenium(II) complex, 30-32% Ru |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only