accessibility menu, dialog, popup

UserName

RuCH2H2CCH2H2CCH3CH3

CAS RN 12289-94-0

IUPAC Name: Bis(2-methylallyl)(1,5-cyclooctadiene)ruthenium(II)
Synonyms: —
Molecular Weight (g/mol): 319.45
Molecular Formula: C16H26Ru
InChi Key: —
SMILES: [Ru++].Cc(:c):c.Cc(:c):c.C1C\C=C/CC\C=C/1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results