missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) Ether, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor B10991LTGL

 View more versions of this product

Catalog No. 18609081


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

111-96-6
Amber Glass Bottle
1 L
COCCOCCOC
134.18
Reagent
1
C6H14O3
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
0.99

Chemical Identifiers

111-96-6
134.18
1-methoxy-2-(2-methoxyethoxy)ethane
C6H14O3
SBZXBUIDTXKZTM-UHFFFAOYSA-N
COCCOCCOC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.