Learn More
Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™
CAS: 111-96-6 | C6H14O3 | 134.18 g/mol
$110.25 - $110.25
Chemical Identifiers
| CAS | 111-96-6 |
|---|---|
| Molecular Formula | C6H14O3 |
| Molecular Weight (g/mol) | 134.18 |
| InChI Key | SBZXBUIDTXKZTM-UHFFFAOYSA-N |
| Synonym | diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane |
| PubChem CID | 8150 |
| ChEBI | CHEBI:46784 |
| IUPAC Name | 1-methoxy-2-(2-methoxyethoxy)ethane |
| SMILES | COCCOCCOC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC443101000
|
Thermo Scientific Chemicals
443101000 |
100 mL |
Each for $110.25
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111-96-6 | |
| 134.18 | |
| diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane | |
| CHEBI:46784 | |
| COCCOCCOC |
| C6H14O3 | |
| SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
| 8150 | |
| 1-methoxy-2-(2-methoxyethoxy)ethane |
Specifications
| 111-96-6 | |
| 0.9370g/mL | |
| 51°C | |
| 99% min. (GC) | |
| 1.4070 to 1.4090 | |
| 100 mL | |
| 15,3184 | |
| Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, hydrocarbon solvents | |
| COCCOCCOC | |
| 134.18 | |
| 2 mPa.s (20°C) | |
| 134.18 | |
| Extra Dry over Molecular Sieve |
| -64.0°C | |
| 162.0°C | |
| Authentic | |
| C6H14O3 | |
| (CH3OCH2CH2)2O | |
| 0.937 | |
| diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane | |
| SBZXBUIDTXKZTM-UHFFFAOYSA-N | |
| 1-methoxy-2-(2-methoxyethoxy)ethane | |
| 8150 | |
| CHEBI:46784 | |
| 99+% | |
| Bis(2-methoxyethyl) ether |
Safety and Handling
GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
GHS Signal Word: Danger
EINECSNumber : 203-924-4
RUO – Research Use Only