missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-methoxyethyl) ether, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 443100010

Catalog No. AC443100010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Bis(2-methoxyethyl) ether
0.9370g/mL
51°C
99% min. (GC)
1.4070 to 1.4090
1L
15,3184
Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, hydrocarbon solvents
COCCOCCOC
134.18
2 mPa.s (20°C)
134.18
Extra Dry over Molecular Sieve
111-96-6
162.0°C
Authentic
C6H14O3
(CH3OCH2CH2)2O
0.937
diglyme, bis 2-methoxyethyl ether, 2-methoxyethyl ether, 1-methoxy-2-2-methoxyethoxy ethane, diethylene glycol dimethyl ether, poly-solv, di 2-methoxyethyl ether, dimethyl carbitol, 2,5,8-trioxanonane
SBZXBUIDTXKZTM-UHFFFAOYSA-N
1-methoxy-2-(2-methoxyethoxy)ethane
8150
CHEBI:46784
99+%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May damage fertility or the unborn child.
Flammable liquid and vapor.
May form explosive peroxides.

GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.

GHS Signal Word: Danger

EINECSNumber : 203-924-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only