missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-mercaptoethyl) Ether 95.0+%, TCI America™

Catalog No. p-7134187
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O010625ML 25 mL
O0106500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O010625ML Supplier TCI America Supplier No. O010625ML
Only null left

Chemical Identifiers

CAS 2150-02-9
Molecular Formula C4H10OS2
Molecular Weight (g/mol) 138.24
MDL Number MFCD00004889
InChI Key CNDCQWGRLNGNNO-UHFFFAOYSA-N
PubChem CID 75073
IUPAC Name 2-(2-sulfanylethoxy)ethanethiol
SMILES C(CS)OCCS

Specifications

Color Yellow
Boiling Point 217°C
Quantity 25 mL
Synonym Di(2-mercaptoethyl) Ether, 2-Mercaptoethyl Ether, 2,2′C-Oxydiethanethiol
Formula Weight 138.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bis(2-mercaptoethyl) Ether