accessibility menu, dialog, popup

UserName

HSOSH

CAS RN 2150-02-9

IUPAC Name: 2-(2-sulfanylethoxy)ethane-1-thiol
Synonyms: dithiodiglycol
Molecular Weight (g/mol): 138.24
Molecular Formula: C4H10OS2
InChi Key: CNDCQWGRLNGNNO-UHFFFAOYSA-N
SMILES: SCCOCCS
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results