missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-mercaptoethyl) Ether 95.0+%, TCI America™

Catalog No. O0106500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0106500ML 500 mL
O010625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0106500ML Supplier TCI America Supplier No. O0106500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2150-02-9
Molecular Formula C4H10OS2
Molecular Weight (g/mol) 138.24
MDL Number MFCD00004889
InChI Key CNDCQWGRLNGNNO-UHFFFAOYSA-N
PubChem CID 75073
IUPAC Name 2-(2-sulfanylethoxy)ethanethiol
SMILES C(CS)OCCS

Specifications

Color Yellow
Boiling Point 217°C
Quantity 500 mL
Synonym Di(2-mercaptoethyl) Ether, 2-Mercaptoethyl Ether, 2,2′-Oxydiethanethiol
Formula Weight 138.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bis(2-mercaptoethyl) Ether