missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxyethyl) Terephthalate 85.0+%, TCI America™

Catalog No. B342925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B342925G 25 g
B3429500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B342925G Supplier TCI America Supplier No. B342925G
Only null left

Chemical Identifiers

CAS 959-26-2
Molecular Formula C12H14O6
Molecular Weight (g/mol) 254.238
MDL Number MFCD00081066
InChI Key QPKOBORKPHRBPS-UHFFFAOYSA-N
Synonym Terephthalic Acid Bis(2-hydroxyethyl) Ester, BHET
PubChem CID 13739
IUPAC Name bis(2-hydroxyethyl) benzene-1,4-dicarboxylate
SMILES C1=CC(=CC=C1C(=O)OCCO)C(=O)OCCO

Specifications

Melting Point 106°C
Color White
Quantity 25 g
Formula Weight 254.24
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis(2-hydroxyethyl) Terephthalate
EINECSNumber (3)-1322&(3)-1327
TSCA Yes