missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-hydroxyethyl)dimethylammonium Chloride 98.0+%, TCI America™
Supplier: TCI America B438425G
Specifications
| Bis(2-hydroxyethyl)dimethylammonium Chloride | |
| White | |
| MFCD00041987 | |
| bis 2-hydroxyethyl dimethylammonium chloride, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethylethanaminium chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride 1:1, bis 2-hydroxyethyl dimethylazanium chloride, bis 2-hydroxyethyl-dimethylazanium chloride, n-2-hydroxyethyl-n,n-dimethyl-n-2-hydroxyethyl ammonium chloride | |
| C[N+](C)(CCO)CCO.[Cl-] | |
| 169.649 | |
| 169.65 | |
| Crystalline Powder |
| 38402-02-7 | |
| C6H16ClNO2 | |
| 25 g | |
| OPVXPOIGBSHXNM-UHFFFAOYSA-M | |
| bis(2-hydroxyethyl)-dimethylazanium;chloride | |
| 162303 | |
| ≥98.0% (T) |
Chemical Identifiers
| 38402-02-7 | |
| 169.649 | |
| OPVXPOIGBSHXNM-UHFFFAOYSA-M | |
| 162303 | |
| C[N+](C)(CCO)CCO.[Cl-] |
| C6H16ClNO2 | |
| MFCD00041987 | |
| bis 2-hydroxyethyl dimethylammonium chloride, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethylethanaminium chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride, ethanaminium, 2-hydroxy-n-2-hydroxyethyl-n,n-dimethyl-, chloride 1:1, bis 2-hydroxyethyl dimethylazanium chloride, bis 2-hydroxyethyl-dimethylazanium chloride, n-2-hydroxyethyl-n,n-dimethyl-n-2-hydroxyethyl ammonium chloride | |
| bis(2-hydroxyethyl)-dimethylazanium;chloride |
Safety and Handling
TSCA : Yes