missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxy-5-chlorophenyl) Sulfide 98.0+%, TCI America™

Supplier:  TCI America B085025G

Catalog No. B085025G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis(2-hydroxy-5-chlorophenyl) Sulfide
175°C
C12H8Cl2O2S
25 g
ANUSOIHIIPAHJV-UHFFFAOYSA-N
4-chloro-2-(5-chloro-2-hydroxyphenyl)sulfanylphenol
7329
287.15
Crystalline Powder
97-24-5
White-Yellow
MFCD00031479
2,2′C-Dihydroxy-5,5′C-dichlorodiphenyl Sulfide, Bis(5-chloro-2-hydroxyphenyl) Sulfide, 2,2′C-Thiobis(4-chlorophenol)
C1=CC(=C(C=C1Cl)SC2=C(C=CC(=C2)Cl)O)O
287.154
CHEBI:556580
≥98.0% (GC,T)

Chemical Identifiers

97-24-5
287.154
ANUSOIHIIPAHJV-UHFFFAOYSA-N
7329
4-chloro-2-(5-chloro-2-hydroxyphenyl)sulfanylphenol
C12H8Cl2O2S
MFCD00031479
2,2′C-Dihydroxy-5,5′C-dichlorodiphenyl Sulfide, Bis(5-chloro-2-hydroxyphenyl) Sulfide, 2,2′C-Thiobis(4-chlorophenol)
CHEBI:556580
C1=CC(=C(C=C1Cl)SC2=C(C=CC(=C2)Cl)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : SN0350000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions