missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Sulfosuccinate Sodium Salt 95.0+%, TCI America™

$47.46 - $131.00

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym Dioctyl Sulfosuccinate Sodium Salt, Sulfosuccinic Acid Bis(2-ethylhexyl) Ester Sodium Salt, Docusate Sodium Salt
PubChem CID 23673837
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S013925G
View Documents Promotion Details
N/A 25 g
Each for $47.46
Only null left
 
S0139500G
View Documents Promotion Details
N/A 500 g
Each for $131.00
Only null left
 
Description

Description

Specifications

Chemical Identifiers

577-11-7
444.56
APSBXTVYXVQYAB-UHFFFAOYNA-M
23673837
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
C20H37NaO7S
MFCD00012455
Dioctyl Sulfosuccinate Sodium Salt, Sulfosuccinic Acid Bis(2-ethylhexyl) Ester Sodium Salt, Docusate Sodium Salt
sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate

Specifications

Undesignated
Spongy Solid-Wax at 20°C
C20H37NaO7S
Dioctyl Sulfosuccinate Sodium Salt, Sulfosuccinic Acid Bis(2-ethylhexyl) Ester Sodium Salt, Docusate Sodium Salt
[Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
444.56
444.56
Bis(2-ethylhexyl) Sulfosuccinate Sodium Salt
577-11-7
179°C
MFCD00012455
APSBXTVYXVQYAB-UHFFFAOYNA-M
sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
23673837
≥95.0% (T)
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1623

RTECSNumber : WN0525000

TSCA : Yes

SDS
Documents

Documents