missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Sulfosuccinate Sodium Salt 95.0+%, TCI America™

Catalog No. S0139500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0139500G 500 g
S013925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0139500G Supplier TCI America Supplier No. S0139500G

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym Dioctyl Sulfosuccinate Sodium Salt, Sulfosuccinic Acid Bis(2-ethylhexyl) Ester Sodium Salt, Docusate Sodium Salt
PubChem CID 23673837
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Color Undesignated
Physical Form Spongy Solid-Wax at 20°C
Melting Point 179°C
Quantity 500 g
Formula Weight 444.56
Percent Purity ≥95.0% (T)
Chemical Name or Material Bis(2-ethylhexyl) Sulfosuccinate Sodium Salt
EINECSNumber (2)-1623
RTECSNumber WN0525000
TSCA Yes