missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Sebacate 98.0+%, TCI America™
Supplier: TCI America S002525ML
Specifications
| Bis(2-ethylhexyl) Sebacate | |
| -67°C | |
| 377°C | |
| MFCD00009497 | |
| bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate | |
| CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC | |
| 426.68 | |
| 426.68 | |
| Liquid |
| 122-62-3 | |
| Colorless | |
| C26H50O4 | |
| 25 mL | |
| VJHINFRRDQUWOJ-UHFFFAOYNA-N | |
| 1,10-bis(2-ethylhexyl) decanedioate | |
| 31218 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 122-62-3 | |
| 426.68 | |
| VJHINFRRDQUWOJ-UHFFFAOYNA-N | |
| 31218 | |
| CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC |
| C26H50O4 | |
| MFCD00009497 | |
| bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate | |
| 1,10-bis(2-ethylhexyl) decanedioate |
Safety and Handling
EINECSNumber : (2)-0879&(2)-2569
RTECSNumber : VS1000000
TSCA : Yes