missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Sebacate 98.0+%, TCI America™
$37.29 - $67.11
Chemical Identifiers
| CAS | 122-62-3 |
|---|---|
| Molecular Formula | C26H50O4 |
| Molecular Weight (g/mol) | 426.68 |
| MDL Number | MFCD00009497 |
| InChI Key | VJHINFRRDQUWOJ-UHFFFAOYNA-N |
| Synonym | bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate |
| PubChem CID | 31218 |
| IUPAC Name | 1,10-bis(2-ethylhexyl) decanedioate |
| SMILES | CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
S002525ML
|
TCI America
S002525ML |
25 mL |
Each for $37.29
|
|
|||||
|
S0025500ML
|
TCI America
S0025500ML |
500 mL |
Each for $67.11
|
|
|||||
Chemical Identifiers
| 122-62-3 | |
| 426.68 | |
| VJHINFRRDQUWOJ-UHFFFAOYNA-N | |
| 31218 | |
| CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC |
| C26H50O4 | |
| MFCD00009497 | |
| bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate | |
| 1,10-bis(2-ethylhexyl) decanedioate |
Specifications
| 122-62-3 | |
| Colorless | |
| C26H50O4 | |
| 25 mL | |
| VJHINFRRDQUWOJ-UHFFFAOYNA-N | |
| 1,10-bis(2-ethylhexyl) decanedioate | |
| 31218 | |
| ≥98.0% (GC) | |
| Bis(2-ethylhexyl) Sebacate |
| -67°C | |
| 377°C | |
| MFCD00009497 | |
| bis 2-ethylhexyl sebacate, bisoflex, bis 2-ethylhexyl decanedioate, plexol, bisoflex dos, monoplex dos, octoil s, reolube dos, staflex dos, di 2-ethylhexyl sebacate | |
| CCCCC(CC)COC(=O)CCCCCCCCC(=O)OCC(CC)CCCC | |
| 426.68 | |
| 426.68 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0879&(2)-2569
RTECSNumber : VS1000000
TSCA : Yes