missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Phthalate 98.0+%, TCI America™

Catalog No. P0297500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0297500G 500 g
P029725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0297500G Supplier TCI America Supplier No. P0297500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 117-81-7
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00009493
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
Synonym dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil
PubChem CID 8343
ChEBI CHEBI:17747
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Melting Point -50°C
Color Yellow
Boiling Point 361°C
Quantity 500 g
UN Number 3082
Formula Weight 390.56
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Phthalate
EINECSNumber (3)-1307
RTECSNumber TI0350000
TSCA Yes