missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) phthalate, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111019089
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019089 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019089 Supplier Merck Emd Millipore Supplier No. 67261100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034 (ISO Guide 34). This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.

Specifications

Packaging Bottomless Glass Bottle with Fused Cone
CAS 117-81-7
Product Line TraceCERT
Density 0.985 g/mL (at 20°C)
Physical Form Neat
Linear Formula C24H38O4
MDL Number MFCD00009493
Boiling Point 386°C
Synonym DEHP; Phthalic acid bis(2-ethylhexyl ester); 'Dioctyl' phthalate
Quantity 100 mg
Formula Weight 390.56
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC