missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) phthalate, 98+%, Thermo Scientific Chemicals

Catalog No. p-7080752
Change view
Click to view available options
Quantity:
10,000 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1041536 500 g
AAA104150E 2500 g
AAA104150C 10,000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1041536 Supplier Thermo Scientific Chemicals Supplier No. A1041536
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(2-ethylhexyl) phthalate is used as dielectric fluid in capacitors, erasable inks and vacuum pump oils. It is also used as plasticizer. Further, it is used as a hydraulic fluid and as a solvent in glow sticks. In addition to this, it is used to give softness and flexibility to polyvinyl chloride products. It finds application in products like tablecloths, medical tubing, floor tiles, toys, shower curtains, rainwear, dolls, shoes, garden hoses, furniture upholstery, and swimming pool liners.

Solubility
Miscible with oil. Immiscible with water.

Notes
Store in a cool place. Incompatible with nitrates and strong oxidizing agents.

Chemical Identifiers

CAS 117-81-7
Molecular Formula C24H38O4
Molecular Weight (g/mol) 390.564
MDL Number MFCD00009493
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
Synonym dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil
PubChem CID 8343
ChEBI CHEBI:17747
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC

Specifications

Melting Point -55°C
Density 0.985
Boiling Point 230°C (5 mmHg)
Flash Point 207°C (404°F)
Odor Odorless
Refractive Index 1.487
Quantity 500 g
Beilstein 1890696
Merck Index 14,2864
Solubility Information Miscible with oil. Immiscible with water.
Formula Weight 390.56
Percent Purity ≥98%
Chemical Name or Material Bis (2-ethylhexyl) phthalate
Show More Show Less
Hazard Category H350-H360FD
Hazard Statement GHS H Statement
H360
May damage fertility or the unborn child.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 204-211-0
RTECSNumber TI0350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only