missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Maleate 93.0+%, TCI America™

$55.92 - $111.84

Chemical Identifiers

CAS 142-16-5
Molecular Formula C20H36O4
Molecular Weight (g/mol) 340.504
MDL Number MFCD00027250
InChI Key ROPXFXOUUANXRR-YPKPFQOOSA-N
Synonym bis 2-ethylhexyl maleate, di-2-ethylhexyl maleate, rc comonomer dom, di 2-ethylhexyl maleate, maleic acid, bis 2-ethylhexyl ester, 2-butenedioic acid 2z-, bis 2-ethylhexyl ester, maleic acid dioctyl ester, 2-butenedioic acid z-, bis 2-ethylhexyl ester
PubChem CID 5365125
IUPAC Name bis(2-ethylhexyl) (Z)-but-2-enedioate
SMILES CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M001125ML
View Documents
TCI America
M001125ML
25 mL
Each for $55.92
Only null left
 
M0011500ML
View Documents
TCI America
M0011500ML
500 mL
Each for $111.84
Only null left
 
Description

Description

Specifications

Chemical Identifiers

142-16-5
340.504
ROPXFXOUUANXRR-YPKPFQOOSA-N
5365125
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC
C20H36O4
MFCD00027250
bis 2-ethylhexyl maleate, di-2-ethylhexyl maleate, rc comonomer dom, di 2-ethylhexyl maleate, maleic acid, bis 2-ethylhexyl ester, 2-butenedioic acid 2z-, bis 2-ethylhexyl ester, maleic acid dioctyl ester, 2-butenedioic acid z-, bis 2-ethylhexyl ester
bis(2-ethylhexyl) (Z)-but-2-enedioate

Specifications

142-16-5
Colorless
C20H36O4
25 mL
ROPXFXOUUANXRR-YPKPFQOOSA-N
bis(2-ethylhexyl) (Z)-but-2-enedioate
5365125
≥93.0% (GC)
Bis(2-ethylhexyl) Maleate
-60°C
173°C
MFCD00027250
bis 2-ethylhexyl maleate, di-2-ethylhexyl maleate, rc comonomer dom, di 2-ethylhexyl maleate, maleic acid, bis 2-ethylhexyl ester, 2-butenedioic acid 2z-, bis 2-ethylhexyl ester, maleic acid dioctyl ester, 2-butenedioic acid z-, bis 2-ethylhexyl ester
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC
340.504
340.50
Liquid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1107

RTECSNumber : ON0160000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'