missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Maleate 93.0+%, TCI America™

Supplier:  TCI America M0011500ML

 View more versions of this product

Catalog No. M0011500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Bis(2-ethylhexyl) Maleate
-60°C
173°C
MFCD00027250
bis 2-ethylhexyl maleate, di-2-ethylhexyl maleate, rc comonomer dom, di 2-ethylhexyl maleate, maleic acid, bis 2-ethylhexyl ester, 2-butenedioic acid 2z-, bis 2-ethylhexyl ester, maleic acid dioctyl ester, 2-butenedioic acid z-, bis 2-ethylhexyl ester
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC
340.504
340.50
Liquid
142-16-5
Colorless
C20H36O4
500 mL
ROPXFXOUUANXRR-YPKPFQOOSA-N
bis(2-ethylhexyl) (Z)-but-2-enedioate
5365125
≥93.0% (GC)

Chemical Identifiers

142-16-5
340.504
ROPXFXOUUANXRR-YPKPFQOOSA-N
5365125
CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC
C20H36O4
MFCD00027250
bis 2-ethylhexyl maleate, di-2-ethylhexyl maleate, rc comonomer dom, di 2-ethylhexyl maleate, maleic acid, bis 2-ethylhexyl ester, 2-butenedioic acid 2z-, bis 2-ethylhexyl ester, maleic acid dioctyl ester, 2-butenedioic acid z-, bis 2-ethylhexyl ester
bis(2-ethylhexyl) (Z)-but-2-enedioate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1107

RTECSNumber : ON0160000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions