missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Itaconate (stabilized with HQ) 94.0+%, TCI America™

Catalog No. I020525ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I020525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I020525ML Supplier TCI America Supplier No. I020525ML
Only null left

Chemical Identifiers

CAS 2287-83-4
Molecular Formula C21H38O4
Molecular Weight (g/mol) 354.53
MDL Number MFCD00059429
InChI Key CGNRQCGWXXLTIA-UHFFFAOYNA-N
Synonym Dioctyl Itaconate, Itaconic Acid Dioctyl Ester, Itaconic Acid Bis(2-ethylhexyl) Ester
PubChem CID 261108
IUPAC Name 1,4-bis(2-ethylhexyl) 2-methylidenebutanedioate
SMILES CCCCC(CC)COC(=O)CC(=C)C(=O)OCC(CC)CCCC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 354.53
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Itaconate (stabilized with HQ)
EINECSNumber (2)-1130
TSCA No