missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Isophthalate 98.0+%, TCI America™
$65.46 - $98.18
Chemical Identifiers
| CAS | 137-89-3 |
|---|---|
| Molecular Formula | C24H38O4 |
| Molecular Weight (g/mol) | 390.564 |
| MDL Number | MFCD00152284 |
| InChI Key | WXZOXVVKILCOPG-UHFFFAOYSA-N |
| Synonym | Isophthalic Acid Bis(2-ethylhexyl) Ester |
| PubChem CID | 8733 |
| IUPAC Name | bis(2-ethylhexyl) benzene-1,3-dicarboxylate |
| SMILES | CCCCC(CC)COC(=O)C1=CC(=CC=C1)C(=O)OCC(CC)CCCC |
Chemical Identifiers
| 137-89-3 | |
| 390.564 | |
| WXZOXVVKILCOPG-UHFFFAOYSA-N | |
| 8733 | |
| CCCCC(CC)COC(=O)C1=CC(=CC=C1)C(=O)OCC(CC)CCCC |
| C24H38O4 | |
| MFCD00152284 | |
| Isophthalic Acid Bis(2-ethylhexyl) Ester | |
| bis(2-ethylhexyl) benzene-1,3-dicarboxylate |
Specifications
| 137-89-3 | |
| 211°C | |
| MFCD00152284 | |
| Isophthalic Acid Bis(2-ethylhexyl) Ester | |
| CCCCC(CC)COC(=O)C1=CC(=CC=C1)C(=O)OCC(CC)CCCC | |
| 390.564 | |
| 390.56 | |
| Liquid |
| Colorless | |
| C24H38O4 | |
| 25 g | |
| WXZOXVVKILCOPG-UHFFFAOYSA-N | |
| bis(2-ethylhexyl) benzene-1,3-dicarboxylate | |
| 8733 | |
| ≥98.0% (GC) | |
| Bis(2-ethylhexyl) Isophthalate |
Safety and Handling
EINECSNumber : (3)-1319
RTECSNumber : NT2050000
TSCA : Yes