missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Fumarate 98.0+%, TCI America™

Catalog No. p-7129500
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
F011725ML 25 mL
F0117500ML 500 mL
2 options

Catalog No. F011725ML

Supplier: TCI America F011725ML

Only null left

Chemical Identifiers

CAS 141-02-6
Molecular Formula C20H36O4
Molecular Weight (g/mol) 340.504
MDL Number MFCD00065144
InChI Key ROPXFXOUUANXRR-BUHFOSPRSA-N
Synonym Fumaric Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Fumarate, Dioctyl Fumarate, Fumaric Acid Dioctyl Ester
PubChem CID 5370325
IUPAC Name bis(2-ethylhexyl) (E)-but-2-enedioate
SMILES CCCCC(CC)COC(=O)C=CC(=O)OCC(CC)CCCC

Specifications

Color Colorless
Boiling Point 185°C
Quantity 25 mL
Formula Weight 340.50
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Fumarate
EINECSNumber (2)-1099
RTECSNumber LT0525000
TSCA Yes