missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(2-ethylhexyl) Dodecanedioate 94.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 267°C |
| Quantity | 500 mL |
| Formula Weight | 454.74 |
| Percent Purity | ≥94.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Bis(2-ethylhexyl) Dodecanedioate |
Safety and Handling
| TSCA | Yes |