accessibility menu, dialog, popup

UserName

H3CCH3OOOOCH3CH3

CAS RN 19074-24-9

IUPAC Name: 1,12-bis(2-ethylhexyl) dodecanedioate
Synonyms: 1,12-bis(2-ethylhexyl) dodecanedioate
Molecular Weight (g/mol): 454.74
Molecular Formula: C28H54O4
InChi Key: MTPSFISGWJGUGN-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC(=O)CCCCCCCCCCC(=O)OCC(CC)CCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results