missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) Dodecanedioate 94.0+%, TCI America™

Catalog No. D209225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D209225ML 25 mL
D2092500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D209225ML Supplier TCI America Supplier No. D209225ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19074-24-9
Molecular Formula C28H54O4
Molecular Weight (g/mol) 454.736
MDL Number MFCD00144216
InChI Key MTPSFISGWJGUGN-UHFFFAOYSA-N
Synonym Dodecanedioic Acid Di(2-ethylhexyl) Ester, Di(2-ethylhexyl) Dodecanedioate, Dioctyl Dodecanedioate, Dodecanedioic Acid Dioctyl Ester
PubChem CID 29401
IUPAC Name bis(2-ethylhexyl) dodecanedioate
SMILES CCCCC(CC)COC(=O)CCCCCCCCCCC(=O)OCC(CC)CCCC

Specifications

Color Yellow
Boiling Point 267°C
Quantity 25 mL
Formula Weight 454.74
Percent Purity ≥94.0% (GC)
Physical Form Liquid
Chemical Name or Material Bis(2-ethylhexyl) Dodecanedioate