missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-ethylhexyl) adipate, 99%

CAS: 103-23-1 | C22H42O4 | 370.574 g/mol

Supplier:  Thermo Scientific Chemicals L1019522

 View more versions of this product

Catalog No. AAL1019522


Only null left
Explore available promotions

Description

Description

It is employed as a plasticizer. It is a skin conditioning solvent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a plasticizer. It is a skin conditioning solvent.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

Bis(2-ethylhexyl) adipate
-67°C
166°C to 168°C (1 mmHg)
C22H42O4
MFCD00009496
1803774
Soluble in alcohol. Insoluble in water.
CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
370.574
CHEBI:34675
99%
103-23-1
0.99
196°C (384°F)
1.447
100 g
bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
SAOKZLXYCUGLFA-UHFFFAOYSA-N
bis(2-ethylhexyl) hexanedioate
7641
370.58

Chemical Identifiers

103-23-1
370.574
SAOKZLXYCUGLFA-UHFFFAOYSA-N
7641
bis(2-ethylhexyl) hexanedioate
C22H42O4
MFCD00009496
bis 2-ethylhexyl adipate, di 2-ethylhexyl adipate, diethylhexyl adipate, deha, plastomoll doa, beha, vestinol oa, bisoflex doa, effomoll doa
CHEBI:34675
CCCCC(CC)COC(=O)CCCCC(=O)OCC(CC)CCCC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P260-P314-P501c

H373

EINECSNumber : 203-090-1

RTECSNumber : AU9700000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only